α-Naphtholbenzein
α-Naphtholbenzein
α-Naphtholbenzein
Thermo Scientific Chemicals
α-Naphtholbenzein
Indicator
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number ACR415320250
Price (MYR)
916.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
916.00
Add to cart
Chemical Identifiers
CAS919-31-3
IUPAC Name3-(triethoxysilyl)propanenitrile
Molecular FormulaC9H19NO3Si
InChI KeyGBQYMXVQHATSCC-UHFFFAOYSA-N
SMILESCCO[Si](CCC#N)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow