Dimethyl trisulfide, 98+%, Thermo Scientific Chemicals
Dimethyl trisulfide, 98+%, Thermo Scientific Chemicals
Dimethyl trisulfide, 98+%, Thermo Scientific Chemicals
Dimethyl trisulfide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl trisulfide, 98+%, Thermo Scientific Chemicals
CAS: 3658-80-8 | C2H6S3 | 126.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR415030050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDimethyl trisulfide
Name Note98+%
CAS3658-80-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.