Methylsulfuric acid sodium salt, 99%, pract., Thermo Scientific Chemicals
Methylsulfuric acid sodium salt, 99%, pract., Thermo Scientific Chemicals
Methylsulfuric acid sodium salt, 99%, pract., Thermo Scientific Chemicals
Methylsulfuric acid sodium salt, 99%, pract., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methylsulfuric acid sodium salt, 99%, pract., Thermo Scientific Chemicals
CAS: 512-42-5 | CH3NaO4S | 134.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR414900250
Price (MYR)
1,235.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,235.00
Add to cart
Specifications
Chemical Name or MaterialMethylsulfuric acid sodium salt
CAS512-42-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0