L-alpha-Lecithin, granular, from soybean oil
L-alpha-Lecithin, granular, from soybean oil
L-alpha-Lecithin, granular, from soybean oil
L-alpha-Lecithin, granular, from soybean oil
Thermo Scientific Chemicals
L-alpha-Lecithin, granular, from soybean oil
L-alpha-Lecithin, >-97% (phospholipids) min. (as acetone insoluble), C42H80NO8P, CAS Number-8002-43-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number ACR413102500
Price (MYR)
854.00
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
854.00
Add to cart
Specifications
Chemical Name or MaterialL-α-Lecithin
Name NoteSource: Soybean
Acidity35mg KOH/g max.
CAS8002-43-5
Storage Note 1Air sensitive