2-Imidazolidone, 96%, anhydrous, Thermo Scientific Chemicals
2-Imidazolidone, 96%, anhydrous, Thermo Scientific Chemicals
2-Imidazolidone, 96%, anhydrous, Thermo Scientific Chemicals
2-Imidazolidone, 96%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Imidazolidone, 96%, anhydrous, Thermo Scientific Chemicals
CAS: 120-93-4 | C3H6N2O | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number ACR412272500
Price (MYR)
931.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
931.00
Add to cart
Chemical Identifiers
CAS98198-48-2
IUPAC Name2-amino-5-bromo-4-methylpyridin-1-ium
Molecular FormulaC6H8BrN2
InChI KeyJDNCMHOKWINDKI-UHFFFAOYSA-O
SMILESCC1=CC(N)=[NH+]C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)> 96.0%
Appearance (Color)Very pale cream to light brown