(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
CAS: 36567-72-3 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR412120010
Price (MYR)
1,215.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,215.00
Add to cart
Chemical Identifiers
CAS1585-17-7
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White
Melting Point (clear melt)103.0-110.0?C