Acetoin, 96%, dimer, Thermo Scientific Chemicals
Acetoin, 96%, dimer, Thermo Scientific Chemicals
Acetoin, 96%, dimer, Thermo Scientific Chemicals
Acetoin, 96%, dimer, Thermo Scientific Chemicals
Acetoin, 96%, dimer, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetoin, 96%, dimer, Thermo Scientific Chemicals
CAS: 23147-57-1 | C8H16O4 | 176.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR411951000
Price (MYR)
686.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
686.00
Add to cart
Specifications
Chemical Name or MaterialAcetoin
Name Notepract., 93%
CAS23147-57-1
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning