alpha-Hydroxyacetophenone, 97%
alpha-Hydroxyacetophenone, 97%
alpha-Hydroxyacetophenone, 97%
alpha-Hydroxyacetophenone, 97%
Thermo Scientific Chemicals
alpha-Hydroxyacetophenone, 97%
CAS: 582-24-1 | C8H8O2 | 136.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR41190025025 gGlass bottle
Catalog number ACR411900250
Price (MYR)
1,090.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,090.00
Add to cart
Chemical Identifiers
CAS109-66-0
IUPAC Namepentane
Molecular FormulaC5H12
InChI KeyOFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILESCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Color (APHA)≤10
Assay (GC)≥99.0% as n-Pentane
Water Content (Karl Fischer Titration)≤0.02%