n-Heptane, 99+%, for spectroscopy, Thermo Scientific Chemicals
n-Heptane, 99+%, for spectroscopy, Thermo Scientific Chemicals
n-Heptane, 99+%, for spectroscopy, Thermo Scientific Chemicals
n-Heptane, 99+%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Heptane, 99+%, for spectroscopy, Thermo Scientific Chemicals
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
View all
Glass Bottle
Catalog number ACR411250025
Price (MYR)
1,690.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,690.00
Add to cart
Chemical Identifiers
CAS2956-58-3
IUPAC NameN-[2-(prop-2-enamido)ethyl]prop-2-enamide
Molecular FormulaC8H12N2O2
InChI KeyAYGYHGXUJBFUJU-UHFFFAOYSA-N
SMILESC=CC(=O)NCCNC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Polymer Test10% in Methanol: Clear Solution
Assay (HPLC)≥95.0%
Appearance (Color)White to pale cream
For UV/VIS spectroscopy
RUO – Research Use Only