Heparin sodium, Thermo Scientific Chemicals
Heparin sodium, Thermo Scientific Chemicals
Heparin sodium, Thermo Scientific Chemicals
Heparin sodium, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Heparin sodium, Thermo Scientific Chemicals
Heparin Sodium Salt, CAS # 9041-08-1, is derived from porcine intestinal mucosa and shows blood anti-coagulant activity.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Packaging:
View all
Glass Bottle
Catalog number ACR411210010
Price (MYR)
1,092.00
Add to cart
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,092.00
Add to cart
Chemical Identifiers
CAS175135-74-7
IUPAC Name(3-chloro-4-fluorophenyl)hydrazine
Molecular FormulaC6H6ClFN2
InChI KeyDUORBNTWCCJANU-UHFFFAOYSA-N
SMILESNNC1=CC=C(F)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale brown or pale pink
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
FormPowder
General Description
  • Heparin sodium salt displays anti-coagulant activity. It can also efficiently inhibit mesenchymal cell proliferation and promotes the proliferation of primary epithelial cells
  • This compound can bind to antithrombin III and inhibit coagulation proteases
Applications
  • In in vitro studies, it is used as the main component of serum-free media to prevent cell aggregation
  • Heparin sodium salt can also induce the growth of human embryonic stem cells in a defined serum-free medium, acting as a cofactor for a basic fibroblast growth factor (FGF-2)
  • It is also applied in laboratory studies related to stem cell differentiation and growth
  • In in vitro studies this compound exhibits cytotoxic action inducing cancer cell death by interfering with transcription factor activity
RUO – Research Use Only