Glycolic acid 70% in water, Thermo Scientific Chemicals
Glycolic acid 70% in water, Thermo Scientific Chemicals
Glycolic acid 70% in water, Thermo Scientific Chemicals
Glycolic acid 70% in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycolic acid 70% in water, Thermo Scientific Chemicals
CAS: 79-14-1 | C2H4O3 | 76.051 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number ACR411032500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9004-57-3
IUPAC Name2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
Molecular FormulaC23H24N6O4
InChI KeyARSKJXYLLONUAJ-UHFFFAOYSA-N
SMILESCCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or granules
Appearance (Color)White to cream to yellow to pale brown
Assay (unspecified)44.0-51.0% ethoxy groups
Viscosity8-12 cP (5% in 80/20 toluene/ethanol, 25?C)