Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Plastic bottle
Catalog number ACR410775000
Price (MYR)
619.00
Add to cart
Quantity:
500 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
619.00
Add to cart
Specifications
Chemical Name or MaterialFormic acid
CAS64-18-6
Melting Point8.0°C
Boiling Point101.0°C
Health Hazard 1Danger