Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
Plastic bottle
Catalog number ACR410770025
Price (MYR)
1,352.00
Add to cart
Quantity:
2.5 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,352.00
Add to cart
Chemical Identifiers
CAS1003-29-8
IUPAC Name1H-pyrrole-2-carbaldehyde
Molecular FormulaC5H5NO
InChI KeyZSKGQVFRTSEPJT-UHFFFAOYSA-N
SMILESO=CC1=CC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)Low melting crystalline solid
AppearanceMay darken on storage
Infrared spectrumConforms
Melting point40°C to 46°C