Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Thermo Scientific Chemicals
Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Formaldehyde, formalin, CAS # 50-00-0, is a colorless volatile used as a histologic fixative and disinfectant.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number ACR410730050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS123-11-5
IUPAC Name4-methoxybenzaldehyde
Molecular FormulaC8H8O2
InChI KeyZRSNZINYAWTAHE-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Free acid (titration)≤1.0%
Refractive Index1.5710-1.5750 @ 20?C
View more
General Description
  • Formaldehyde (formalin) is an organic compound used for crosslinking proteins in biological samples such as tissue sections, culture cells, and whole specimens
  • Formaldehyde is an intermediate in the combustion of methane and can exist in many forms
  • It is a volatile, colorless liquid; can be stored long-term in appropriate containers
  • Comes stabilized in solution with methanol
Applications
  • May be used in pain research for induction of pain and inflammation
  • Monomeric formaldehyde is formulated especially for laboratory applications
  • Commonly used in RNA isolation as a denaturant
RUO – Research Use Only
General References:
  1. Ramanathan, M.; Porter, D. F.; Khavari P. A. Methods to study RNA-protein interactions. Nat Methods. 2019, 16(3), 225-234. Erratum in: Nat Methods. 2019, 16(4), 351.