Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
CAS: 16961-83-4 | F6H2Si | 144.091 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR410715000
Price (MYR)
523.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
523.00
Add to cart
Chemical Identifiers
CAS111-30-8
IUPAC Namepentanedial
Molecular FormulaC5H8O2
InChI KeySXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILESO=CCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear solution
Strength23.5 to 26.5 %
pH3.2 to 4.2
Color scale=<50 APHA
Methanol=<0.3 % (GC)