Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
CAS: 16961-83-4 | F6H2Si | 144.091 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic bottle
Catalog number ACR410710025
Price (MYR)
1,418.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,418.00
Add to cart
Chemical Identifiers
CAS16961-83-4, 7732-18-5
Specifications
Chemical Name or MaterialFluosilicic acid
ColorColorless
CAS16961-83-4
Health Hazard 1Danger
Density1.2700g/mL