Ethylene glycol, 95%, pure, Thermo Scientific Chemicals
Ethylene glycol, 95%, pure, Thermo Scientific Chemicals
Ethylene glycol, 95%, pure, Thermo Scientific Chemicals
Ethylene glycol, 95%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol, 95%, pure, Thermo Scientific Chemicals
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Glass Bottle
Catalog number ACR410010025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthylene glycol
Melting Point-13.0°C
CAS107-21-1
Health Hazard 1Warning
Density1.1130g/mL