Eosin Y, pure, certified
Eosin Y, pure, certified
Eosin Y, pure, certified
Eosin Y, pure, certified
Thermo Scientific Chemicals
Eosin Y, pure, certified
CAS: 17372-87-1 | C20H12Br4Na2O8 | 745.904 g/mol
Have Questions?
Catalog NumberQuantity
ACR40943025025 g
Catalog number ACR409430250
Price (MYR)
477.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
477.00
Add to cart
Specifications
Chemical Name or MaterialEosin Y
CAS17372-87-1
Health Hazard 1Warning
Absorption Ratio1.21 to 1.77
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
View more
Indicator
RUO – Research Use Only