1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR408820025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS682-30-4
IUPAC Namediethyl ethenylphosphonate
Molecular FormulaC6H13O3P
InChI KeyDREPONDJUKIQLX-UHFFFAOYSA-N
SMILESCCOP(=O)(OCC)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Appearance (Color)Clear, colorless
Phosphorus-31 NMR> 96.0%