Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 95-45-4 | C4H8N2O2 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number ACR408335000
Price (MYR)
1,320.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,320.00
Add to cart
Chemical Identifiers
CAS373-44-4
IUPAC Nameoctane-1,8-bis(aminium) dichloride
Molecular FormulaC8H22Cl2N2
InChI KeyZFLWZOGXFQNIMT-UHFFFAOYSA-N
SMILES[Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point50°C to 54°C
Appearance (Color)White to yellow
Appearance (Form)Fused crystalline mass
Infrared spectrumConforms
Titration with HClO4>=97.5 %