Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 95-45-4 | C4H8N2O2 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Plastic bottle
Catalog number ACR408331000
Price (MYR)
550.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
550.00
Add to cart
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES[H+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Strength0.4 to 0.6 M
Appearance (Form)Solution