N,N-Dimethylcyclohexylamine, 99%, Thermo Scientific Chemicals
N,N-Dimethylcyclohexylamine, 99%, Thermo Scientific Chemicals
N,N-Dimethylcyclohexylamine, 99%, Thermo Scientific Chemicals
N,N-Dimethylcyclohexylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylcyclohexylamine, 99%, Thermo Scientific Chemicals
CAS: 98-94-2 | C8H17N | 127.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number ACR408255000
Price (MYR)
612.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
612.00
Add to cart
Chemical Identifiers
CAS488-47-1
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Cream to brown
Assay (HPLC)≥95.0%