Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Thermo Scientific Chemicals
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
CAS: 140-01-2 | C14H18N3Na5O10 | 503.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
3 kg
Packaging:
Glass bottle
Catalog number ACR407292500
Price (MYR)
436.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
436.00
Add to cart
Chemical Identifiers
CAS140-01-2, 7732-18-5
Specifications
Chemical Name or MaterialDiethylenetriaminepentaacetic acid, pentasodium salt
Name Note40% Aqueous Solution
CAS140-01-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
View more
Chelation/Complexation reagent
RUO – Research Use Only