Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Thermo Scientific Chemicals
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
CAS: 140-01-2 | C14H18N3Na5O10 | 503.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
3 kg
250 g
1 kg
Packaging:
Glass bottle
Catalog number ACR407290030
Price (MYR)
910.00
Add to cart
Quantity:
3 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
910.00
Add to cart
Chemical Identifiers
CAS4394-85-8
IUPAC Namemorpholine-4-carbaldehyde
Molecular FormulaC5H9NO2
InChI KeyLCEDQNDDFOCWGG-UHFFFAOYSA-N
SMILESO=CN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4850-1.4880 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥98.5%
Chelation/Complexation reagent
RUO – Research Use Only