Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR406920025
Price (MYR)
733.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
733.00
Add to cart
Chemical Identifiers
CAS71-30-7
IUPAC Name6-amino-1,2-dihydropyrimidin-2-one
Molecular FormulaC4H5N3O
InChI KeyOPTASPLRGRRNAP-UHFFFAOYSA-N
SMILESNC1=CC=NC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<10 ppm
Sulfated ash=<0.1 % (800°C)
UV spectrophotometry>=98.0 % (lambda max. 275 nm, pH 1)
UV spectrophotometrymolar absorptivity 10400 +/- 2%, on dry basis
Loss on drying=<0.5 % (105°C, 5 hrs)