Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR406920010
Price (MYR)
508.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
508.00
Add to cart
Chemical Identifiers
CAS11006-34-1
IUPAC Namechlorophyllin
Molecular FormulaC34H31CuN4Na3O6
InChI KeyHWDGVJUIHRPKFR-UHFFFAOYNA-I
SMILES[Na+].[Na+].[Na+].[Cu++].CCC1=C(C)C2=N/C/1=CC1=C(C)C(=C([O-])[O-])C(=N1)C(CC([O-])=O)=C1/N=C(C=C3/[N-]/C(=C2)C(C=C)=C3C)C(C)C1CCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to black
Elemental AnalysisCu: 3.5-6.5%
pH9.0-11.0 (1% solution in water)
FormPowder
Loss on Drying≤10.0% (105°C/constant weight)