1,3-Dibromo-2-propanone, 70%, tech.
1,3-Dibromo-2-propanone, 70%, tech.
1,3-Dibromo-2-propanone, 70%, tech.
1,3-Dibromo-2-propanone, 70%, tech.
Thermo Scientific Chemicals
1,3-Dibromo-2-propanone, 70%, tech.
CAS: 816-39-7 | C3H4Br2O | 215.87 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR40655025025 gGlass bottle
Catalog number ACR406550250
Price (MYR)
3,090.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,090.00
Add to cart
Chemical Identifiers
CAS99395-88-7
IUPAC Name(4S)-4-phenyl-1,3-oxazolidin-2-one
Molecular FormulaC9H9NO2
InChI KeyQDMNNMIOWVJVLY-MRVPVSSYSA-N
SMILESO=C1N[C@H](CO1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point130°C to 133°C
Specific optical rotation+48° to +51° (20°C, 589 nm) (c=2, CHCl3)
GC>=98.5 %
Appearance (Color)White