4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
CAS: 81-11-8 | C14H14N2O6S2 | 370.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR406411000
Price (MYR)
508.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
508.00
Add to cart
Specifications
Chemical Name or Material4, 4'-Diaminostilbene-2, 2'-disulfonic acid
Name Note95%
CAS81-11-8
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning