trans-1,4-Diaminocyclohexane, 98+%, Thermo Scientific Chemicals
trans-1,4-Diaminocyclohexane, 98+%, Thermo Scientific Chemicals
trans-1,4-Diaminocyclohexane, 98+%, Thermo Scientific Chemicals
trans-1,4-Diaminocyclohexane, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-1,4-Diaminocyclohexane, 98+%, Thermo Scientific Chemicals
CAS: 2615-25-0 | C6H14N2 | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number ACR406360250
Price (MYR)
1,021.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,021.00
Add to cart
Chemical Identifiers
CAS1544-68-9
IUPAC Name1-fluoro-4-isothiocyanatobenzene
Molecular FormulaC7H4FNS
InChI KeyNFIUJHJMCQQYDL-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.6185-1.6225 @ 20?C
Appearance (Color)Colorless to white to pale cream to pale yellow
FormFused solid or clear liquid as melt