Dextran, High Fraction, Thermo Scientific Chemicals
Dextran, High Fraction, Thermo Scientific Chemicals
Dextran, High Fraction, Thermo Scientific Chemicals
Dextran, High Fraction, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dextran, High Fraction, Thermo Scientific Chemicals
CAS: 9004-54-0 | (C6H9O5)n(C6H10O5)m | 504.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR406261000
Price (MYR)
1,037.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,037.00
Add to cart
Chemical Identifiers
CAS7758-09-0
IUPAC Namepotassium nitrite
Molecular FormulaKNO2
InChI KeyBXNHTSHTPBPRFX-UHFFFAOYSA-M
SMILES[K+].[O-]N=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Arsenic (As)=<1 ppm
Lead (Pb)=<10 ppm
Titration with KMnO4>=96.0 %
Iron (Fe)=<10 ppm