Cyclohexyldiphenylphosphine, 98%, Thermo Scientific Chemicals
Cyclohexyldiphenylphosphine, 98%, Thermo Scientific Chemicals
Cyclohexyldiphenylphosphine, 98%, Thermo Scientific Chemicals
Cyclohexyldiphenylphosphine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexyldiphenylphosphine, 98%, Thermo Scientific Chemicals
CAS: 6372-42-5 | C18H21P | 268.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR406100050
Price (MYR)
1,354.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,354.00
Add to cart
Chemical Identifiers
CAS7240-90-6
IUPAC NameX-gal
Molecular FormulaC23H41Cu2N5O5
InChI KeyGGILAJWFDJWIQO-UHFFFAOYSA-N
SMILES[Cu+].[Cu++].CC(O)O.CC(O)O.CN1CCN(CC2=CC(=CC(CN3CCN(C)CC3)=C2O)C#N)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Optical Rotation-61.6 ± 0.1? (C=1 in DMF:H2O 50:50)
Appearance (Color)White
Assay (HPLC)≥98.0%