Cyclohexanone, 99+%, Thermo Scientific Chemicals
Cyclohexanone, 99+%, Thermo Scientific Chemicals
Cyclohexanone, 99+%, Thermo Scientific Chemicals
Cyclohexanone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexanone, 99+%, Thermo Scientific Chemicals
CAS: 108-94-1 | C6H10O | 98.145 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Catalog number ACR406092500
Price (MYR)
434.00
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
434.00
Add to cart
Specifications
Chemical Name or MaterialCyclohexanone
Melting Point-47.0°C
CAS108-94-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement Flammable liquid and vapor. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes serious eye damage. Causes skin irritation.