β-Cyclodextrin, 98%, Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
CAS: 7585-39-9 | C42H70O35 | 1134.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ACR406005000
Price (MYR)
2,486.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
2,486.00
Add to cart
Specifications
Chemical Name or Materialβ-Cyclodextrin
CAS7585-39-9
Melting Point310.0°C
EINECS Number231-493-2
Loss on Drying14% max. (105°C, 8 hrs)