β-Cyclodextrin, 98%, Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
CAS: 7585-39-9 | C42H70O35 | 1134.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number ACR406001000
Price (MYR)
805.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
805.00
Add to cart
Chemical Identifiers
CAS1155-64-2
IUPAC Name(2S)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoic acid
Molecular FormulaC14H20N2O4
InChI KeyCKGCFBNYQJDIGS-LBPRGKRZSA-N
SMILESN[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
FormPowder
Optical Rotation15.3 ± 0.5? (C=1 in 1N HCl)