Cyanocobalamin, 96%
Cyanocobalamin, 96%
Cyanocobalamin, 96%
Thermo Scientific Chemicals
Cyanocobalamin, 96%
CAS: 68-19-9 | C63H88CoN14O14P | 1355.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR405925000
Price (MYR)
709.00
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
709.00
Add to cart
Chemical Identifiers
CAS6564-72-3
IUPAC Name3,4,5-tris(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-ol
Molecular FormulaC34H36O6
InChI KeyOGOMAWHSXRDAKZ-UHFFFAOYNA-N
SMILESOC1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Optical Rotation49.8 ± 0.1? (C=2 in dioxane)
Melting Point (clear melt)149.0-156.0?C