Cyanocobalamin, 96%
Cyanocobalamin, 96%
Cyanocobalamin, 96%
Thermo Scientific Chemicals
Cyanocobalamin, 96%
CAS: 68-19-9 | C63H88CoN14O14P | 1355.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR405920250
Price (MYR)
8,526.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
8,526.00
Add to cart
Chemical Identifiers
CAS121-01-7
IUPAC Name4-nitro-2-(trifluoromethyl)aniline
Molecular FormulaC7H5F3N2O2
InChI KeyHOTZLWVITTVZGY-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1C(F)(F)F)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)89.0-96.0?C