p-Cresol, 99+%, pure, Thermo Scientific Chemicals
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
CAS: 106-44-5 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ACR405745000
Price (MYR)
686.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
686.00
Add to cart
Chemical Identifiers
CAS1065010-87-8
IUPAC Namepotassium cyclopropyltrifluoroboranuide
Molecular FormulaC3H5BF3K
InChI KeyCFMLURFHOSOXRC-UHFFFAOYSA-N
SMILES[K+].F[B-](F)(F)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Solid
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure