p-Cresol, 99+%, pure, Thermo Scientific Chemicals
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
CAS: 106-44-5 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ACR405741000
Price (MYR)
398.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
398.00
Add to cart
Chemical Identifiers
CAS2039-67-0
IUPAC Namehydrogen 2-(3-methoxyphenyl)ethan-1-amine chloride
Molecular FormulaC9H14ClNO
InChI KeyKDRBQDYHPKRFGX-UHFFFAOYSA-N
SMILES[H+].[Cl-].COC1=CC=CC(CCN)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.0%