o-Cresol, 99%, AcroSeal™, Thermo Scientific Chemicals
o-Cresol, 99%, AcroSeal™, Thermo Scientific Chemicals
o-Cresol, 99%, AcroSeal™, Thermo Scientific Chemicals
o-Cresol, 99%, AcroSeal™, Thermo Scientific Chemicals
o-Cresol, 99%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Cresol, 99%, AcroSeal™, Thermo Scientific Chemicals
CAS: 95-48-7 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACR405731000
Price (MYR)
475.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
475.00
Add to cart
Chemical Identifiers
CAS2546-56-7
SpecificationsSpecification SheetSpecification Sheet
GC>=94.0 %
Appearance (Color)Colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid