Citronellal, 95+%, Thermo Scientific Chemicals
Citronellal, 95+%, Thermo Scientific Chemicals
Citronellal, 95+%, Thermo Scientific Chemicals
Citronellal, 95+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citronellal, 95+%, Thermo Scientific Chemicals
CAS: 106-23-0 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR405291000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12055-62-8
IUPAC Namediholmium(3+) trioxidandiide
Molecular FormulaHo2O3
InChI KeyOWCYYNSBGXMRQN-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Ho+3].[Ho+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.005% max.