3-(Chlorosulfonyl)benzoyl chloride, 98%
3-(Chlorosulfonyl)benzoyl chloride, 98%
3-(Chlorosulfonyl)benzoyl chloride, 98%
3-(Chlorosulfonyl)benzoyl chloride, 98%
Thermo Scientific Chemicals
3-(Chlorosulfonyl)benzoyl chloride, 98%
CAS: 4052-92-0 | C7H4Cl2O3S | 239.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR40507025025 gGlass bottle
Catalog number ACR405070250
Price (MYR)
1,825.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,825.00
Add to cart
Chemical Identifiers
CAS622-40-2
IUPAC Name2-(morpholin-4-yl)ethan-1-ol
Molecular FormulaC6H13NO2
InChI KeyKKFDCBRMNNSAAW-UHFFFAOYSA-N
SMILESOCCN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Water=<0.5 %
Infrared spectrumConforms
Refractive index1.4760 to 1.4780 (20°C, 589 nm)
Appearance (Form)Liquid