5-Chlorosalicylaldehyde, 98%, Thermo Scientific Chemicals
5-Chlorosalicylaldehyde, 98%, Thermo Scientific Chemicals
5-Chlorosalicylaldehyde, 98%, Thermo Scientific Chemicals
5-Chlorosalicylaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Chlorosalicylaldehyde, 98%, Thermo Scientific Chemicals
CAS: 635-93-8 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
View all
Plastic bottle
Catalog number ACR405030250
Price (MYR)
924.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
924.00
Add to cart
Chemical Identifiers
CAS123333-66-4
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96.1% min