5-Chlorosalicylaldehyde, 98%, Thermo Scientific Chemicals
5-Chlorosalicylaldehyde, 98%, Thermo Scientific Chemicals
5-Chlorosalicylaldehyde, 98%, Thermo Scientific Chemicals
5-Chlorosalicylaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Chlorosalicylaldehyde, 98%, Thermo Scientific Chemicals
CAS: 635-93-8 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number ACR405030050
Price (MYR)
436.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
436.00
Add to cart
Chemical Identifiers
CAS86-81-7
IUPAC Name3,4,5-trimethoxybenzaldehyde
Molecular FormulaC10H12O4
InChI KeyOPHQOIGEOHXOGX-UHFFFAOYSA-N
SMILESCOC1=CC(C=O)=CC(OC)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)White to cream
Appearance (Form)Powder or flakes
Melting point73°C to 76°C