4-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
4-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
4-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
4-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
CAS: 1073-70-7 | C6H7ClN2·HCl | 179.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number ACR404950250
Price (MYR)
699.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
699.00
Add to cart
Chemical Identifiers
CAS86-81-7
IUPAC Name3,4,5-trimethoxybenzaldehyde
Molecular FormulaC10H12O4
InChI KeyOPHQOIGEOHXOGX-UHFFFAOYSA-N
SMILESCOC1=CC(C=O)=CC(OC)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Free acid (titration)≤1%
Identification (FTIR)Conforms
Appearance (Color)White to cream to pale orange
Melting Point (clear melt)72.0-78.0?C