2-Chloronicotinic acid, 99%
2-Chloronicotinic acid, 99%
2-Chloronicotinic acid, 99%
2-Chloronicotinic acid, 99%
Thermo Scientific Chemicals
2-Chloronicotinic acid, 99%
CAS: 2942-59-8 | C6H4ClNO2 | 157.56 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR40486050050 gGlass bottle
Catalog number ACR404860500
Price (MYR)
844.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
844.00
Add to cart
Chemical Identifiers
CAS5271-67-0
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5880-1.5920 @ 20?C
Appearance (Color)Clear colorless to pale yellow or brown or green
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥97.5%