2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
CAS: 91-58-7 | C10H7Cl | 162.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR404840250
Price (MYR)
8,183.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
8,183.00
Add to cart
Chemical Identifiers
CAS194491-31-1
IUPAC Nametetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Molecular FormulaC10H12N2Na4O8
InChI KeyUEUXEKPTXMALOB-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to light yellow
Infrared spectrumConforms
Loss on drying5 % to 15 % (160°C)
Titration Complexometric>=97.5 % (on dry substance)