2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
CAS: 91-58-7 | C10H7Cl | 162.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR404840050
Price (MYR)
1,897.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,897.00
Add to cart
Chemical Identifiers
CAS51987-73-6
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%