Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number ACR404415000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS51-79-6
IUPAC Nameethyl carbamate
Molecular FormulaC3H7NO2
InChI KeyJOYRKODLDBILNP-UHFFFAOYSA-N
SMILESCCOC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Proton NMRConforms
Sulfated Ash≤0.1%
Water Content (Karl Fischer Titration)≤0.5%
FormCrystalline powder