Cellulose Acetate Butyrate, Butyryl Content 35-39%
Cellulose Acetate Butyrate, Butyryl Content 35-39%
Cellulose Acetate Butyrate, Butyryl Content 35-39%
Cellulose Acetate Butyrate, Butyryl Content 35-39%
Thermo Scientific Chemicals
Cellulose Acetate Butyrate, Butyryl Content 35-39%
CAS: 9004-36-8 | (C12H14O10)n
Have Questions?
Catalog NumberQuantityPackaging
ACR404255000500 gPlastic bottle
Catalog number ACR404255000
Price (MYR)
911.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
911.00
Add to cart
Chemical Identifiers
CAS123334-20-3
IUPAC Nameoxovanadiumbis(ylium) sulfate
Molecular FormulaO5SV
InChI KeyUUUGYDOQQLOJQA-UHFFFAOYSA-L
SMILESO=[V++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.