Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
1 kg
2.5 kg
Catalog number ACR404165000
Price (MYR)
487.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
487.00
Add to cart
Specifications
Chemical Name or MaterialCastor oil
CAS8001-79-4
Melting Point-10°C
ColorYellow
Density0.9570g/mL